siRNA Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022210.1 | NP_071546.1 | 4 | 222 | ||
XM_006240248.2 | XP_006240310.1 | 3 | 421 | |||
XM_008764846.1 | XP_008763068.1 | 4 | 453 | |||
GenBank mRNA | BC099778.1 | - | 4 | 384 | 7933028 | |
D14447.1 | - | 4 | 222 | |||
D14448.1 | - | 3 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
