siRNA Details
Gene Symbol: Max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022210.1 | NP_071546.1 | 3 | 96 | ||
XM_006240248.2 | XP_006240310.1 | 2 | 295 | |||
XM_008764846.1 | XP_008763068.1 | 3 | 327 | |||
GenBank mRNA | BC099778.1 | - | 3 | 258 | 7933028 | |
D14447.1 | - | 3 | 96 | |||
D14448.1 | - | 2 | 69 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008558, NM_001146176, BC138671, BC138672, BC145369, and others.
Gene Ontology Categories:
