siRNA Details
Gene Symbol: Fut4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022219.3 | NP_071555.3 | 1 | 1044 | ||
GenBank mRNA | AB049938.1 | - | 1 | 689 | ||
BC061776.1 | - | 1 | 1044 | 5621698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
