siRNA Details
Gene Symbol: Actn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031675.2 | NP_113863.2 | 7 | 782 | ||
XM_006228687.1 | XP_006228749.1 | 7 | 821 | |||
XM_006228688.1 | XP_006228750.1 | 7 | 821 | |||
XM_006228689.1 | XP_006228751.1 | 7 | 821 | |||
XM_006228690.2 | XP_006228752.1 | 7 | 1100 | |||
XM_008759185.1 | XP_008757407.1 | 7 | 821 | |||
GenBank mRNA | AF190909.1 | - | 7 | 707 | ||
AY724534.1 | - | 7 | 751 | |||
BC061788.1 | - | 7 | 773 | 5623735 | ||
U19893.1 | - | 7 | 783 |

More Information
Other Species Targeted

Gene Ontology Categories:
