siRNA Details
Gene Symbol: Fxyd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022005.2 | NP_071288.1 | 3 | 306 | ||
XM_008766171.1 | XP_008764393.1 | 3 | 335 | |||
GenBank mRNA | AA799380.1 | - | 3 | 256 | ||
AB030908.1 | - | 3 | 120 | |||
AF142439.1 | - | 3 | 116 | |||
BC072528.1 | - | 3 | 112 | 7096832 | ||
CB741544.1 | - | 3 | 328 |

More Information
Other Species Targeted

Gene Ontology Categories:
