siRNA Details
Gene Symbol: Prlh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022222.1 | NP_071558.1 | 1 | 86 | ||
XM_006245478.2 | XP_006245540.1 | 2 | 790 | |||
GenBank mRNA | AB015418.1 | - | 1 | 86 | ||
AF521930.1 | - | 1 | 86 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
