siRNA Details
Gene Symbol: Hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031682.1 | NP_113870.1 | 6 | 713 | ||
GenBank mRNA | AF049878.1 | - | 6 | 701 | ||
AF069770.1 | - | 6 | 713 | |||
BC158587.1 | - | 6 | 707 | 9089087 | ||
FQ220319.1 | - | 4 | 454 | |||
FQ225948.1 | - | 4 | 454 | |||
FQ228203.1 | - | 4 | 454 | |||
FQ230672.1 | - | 4 | 455 | |||
FQ231826.1 | - | 4 | 454 | |||
FQ232016.1 | - | 4 | 454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
