siRNA Details
Gene Symbol: Lgmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022226.2 | NP_071562.2 | 12, 13 | 1311 | ||
GenBank mRNA | AB032766.1 | - | 12, 13 | 1306 | ||
AF154349.1 | - | 12, 13 | 1311 | |||
BC087708.1 | - | 1348 | 7315308 |
