siRNA Details
Gene Symbol: Asic5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022227.1 | NP_071563.1 | 9 | 1357 | ||
GenBank mRNA | Y19034.1 | - | 9 | 1357 |

More Information
Other Species Targeted

Gene Ontology Categories:
