siRNA Details
Gene Symbol: Nmu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022239.2 | NP_071575.1 | 2 | 268 | ||
XM_006250872.2 | XP_006250934.1 | 2 | 1088 | |||
GenBank mRNA | AJ510132.1 | - | 1 | 27 | ||
M94555.1 | - | 2 | 268 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
