siRNA Details
Gene Symbol: Arhgap17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270692.1 | NP_001257621.1 | 9 | 745 | ||
NM_001270693.1 | NP_001257622.1 | 9 | 745 | |||
NM_001270694.1 | NP_001257623.1 | 9 | 745 | |||
NM_022244.2 | NP_071580.1 | 9 | 745 | |||
XM_006230194.2 | XP_006230256.1 | 9 | 797 | |||
XM_008759791.1 | XP_008758013.1 | 9 | 797 | |||
XM_008759792.1 | XP_008758014.1 | 9 | 798 | |||
XM_008759793.1 | XP_008758015.1 | 9 | 797 | |||
XM_008759794.1 | XP_008758016.1 | 9 | 796 | |||
GenBank mRNA | AB042827.1 | - | 9 | 744 | ||
AB060556.1 | - | 9 | 744 | |||
AB060557.1 | - | 9 | 744 | |||
AB080637.1 | - | 9 | 744 | |||
BC085736.1 | - | 9 | 746 | 7130218 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001122643, NM_001122640, NM_001122641, NM_144529, NM_001122642, and others.
Gene Ontology Categories:
