siRNA Details
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022251.2 | NP_071587.2 | 1 | 877 | ||
GenBank mRNA | AF146518.1 | - | 1 | 239 | ||
AF214568.1 | - | 1 | 694 | |||
BC066663.1 | - | 1 | 699 | 5623429 |
Gene Symbol: LOC100910501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761484.1 | XP_008759706.1 | 1 | 725 |
