siRNA Details
Gene Symbol: Cd244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022259.1 | NP_071595.1 | 2 | 278 | ||
XM_006250259.2 | XP_006250321.1 | 2 | 1657 | |||
GenBank mRNA | AF156988.1 | - | 2 | 338 | ||
AF156989.1 | - | 2 | 278 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
