siRNA Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022260.3 | NP_071596.1 | 1 | 48 | ||
XM_008760546.1 | XP_008758768.1 | 2 | 641 | |||
GenBank mRNA | AF072124.1 | - | 1 | 49 | ||
BC070936.1 | - | 1 | 49 | 7099111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
