siRNA Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022260.3 | NP_071596.1 | 5, 6 | 680 | ||
XM_008760546.1 | XP_008758768.1 | 6, 7 | 1273 | |||
XM_008760547.1 | XP_008758769.1 | 5, 6 | 1655 | |||
XM_008760548.1 | XP_008758770.1 | 5, 6 | 982 | |||
XM_008760549.1 | XP_008758771.1 | 6, 7 | 862 | |||
XM_008760550.1 | XP_008758772.1 | 5, 6 | 790 | |||
GenBank mRNA | AF072124.1 | - | 5, 6 | 681 | ||
BC070936.1 | - | 5, 6 | 681 | 7099111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
