siRNA Details
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022266.2 | NP_071602.1 | 4, 5 | 972 | ||
GenBank mRNA | AB023068.1 | - | 4, 5 | 960 | ||
AF079531.1 | - | 1, 2 | 106 | |||
AF120275.1 | - | 4, 5 | 972 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
