siRNA Details
Gene Symbol: Casp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022277.1 | NP_071613.1 | 8 | 1334 | ||
XM_006245014.2 | XP_006245076.1 | 8 | 1316 | |||
XM_006245015.2 | XP_006245077.1 | 8 | 1457 | |||
XM_008767135.1 | XP_008765357.1 | 8 | 1316 | |||
GenBank mRNA | AF279308.1 | - | 8 | 1334 | ||
AF288372.1 | - | 8 | 1069 | |||
FQ226407.1 | - | 8 | 1348 |
