siRNA Details
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022282.1 | NP_071618.1 | 12, 13 | 1677 | ||
GenBank mRNA | U49049.1 | - | 12, 13 | 1677 |

More Information
Other Species Targeted

Gene Ontology Categories:
