siRNA Details
Gene Symbol: Hapln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022285.1 | NP_071621.1 | 7 | 1354 | ||
XM_006232751.2 | XP_006232813.1 | 7 | 1436 | |||
XM_008761208.1 | XP_008759430.1 | 8 | 1477 | |||
XM_008761209.1 | XP_008759431.1 | 7 | 1662 | |||
GenBank mRNA | AB049056.1 | - | 7 | 1354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
