siRNA Details
Gene Symbol: Slc26a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022287.2 | NP_071623.2 | 3 | 899 | ||
GenBank mRNA | BC085735.1 | - | 3 | 899 | 7108820 | |
L23413.1 | - | 3 | 1221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
