siRNA Details
Gene Symbol: Gosr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031685.2 | NP_113873.1 | 2, 3 | 99 | ||
GenBank mRNA | AF007549.1 | - | 2, 3 | 113 | ||
BC061994.1 | - | 2, 3 | 99 | 5599557 | ||
U91539.1 | - | 2, 3 | 98 |

More Information
Other Species Targeted

Gene Ontology Categories:
