siRNA Details
Gene Symbol: Sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171983.2 | NP_741984.2 | 17 | 2314 | ||
XM_008761673.1 | XP_008759895.1 | 17 | 2314 | |||
XM_008761674.1 | XP_008759896.1 | 17 | 2314 | |||
XM_008761675.1 | XP_008759897.1 | 17 | 2314 | |||
XM_008761676.1 | XP_008759898.1 | 17 | 2314 | |||
XM_008761677.1 | XP_008759899.1 | 17 | 2314 | |||
XM_008761678.1 | XP_008759900.1 | 17 | 2314 | |||
GenBank mRNA | AF084186.1 | - | 16 | 2206 | ||
BC070885.1 | - | 17 | 2278 | 7098134 | ||
GQ502182.2 | - | 16 | 2206 | |||
X90845.1 | - | 17 | 2314 |
