siRNA Details
Gene Symbol: Sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171983.2 | NP_741984.2 | 16 | 2156 | ||
GenBank mRNA | AF084186.1 | - | 15 | 2048 | ||
GQ502182.2 | - | 15 | 2048 | |||
X90845.1 | - | 16 | 2156 |
