siRNA Details
Gene Symbol: Basp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022300.1 | NP_071636.1 | 2 | 708 | ||
GenBank mRNA | D14441.1 | - | 2 | 708 |

More Information
Other Species Targeted

Gene Ontology Categories:
