siRNA Details
Gene Symbol: Mafg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022386.2 | NP_071781.1 | 3 | 334 | ||
XM_006247912.2 | XP_006247974.1 | 3 | 4417 | |||
XM_006247913.2 | XP_006247975.1 | 4 | 3418 | |||
XM_006247914.2 | XP_006247976.1 | 2 | 337 | |||
XM_006247916.2 | XP_006247978.1 | 2 | 2114 | |||
XM_006247917.2 | XP_006247979.1 | 2 | 4886 | |||
XM_006247918.2 | XP_006247980.1 | 2 | 1497 | |||
XM_006247919.2 | XP_006247981.1 | 2 | 2044 | |||
XM_006247921.2 | XP_006247983.1 | 3 | 1095 | |||
XM_006247922.2 | XP_006247984.1 | 3 | 256 | |||
XM_006247923.2 | XP_006247985.1 | 3 | 1025 | |||
XM_008768482.1 | XP_008766704.1 | 3 | 3867 | |||
GenBank mRNA | AB026487.1 | - | 2 | 208 | ||
AB050011.1 | - | 1 | 261 | |||
BC078828.1 | - | 3 | 334 | 7127046 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
