siRNA Details
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022389.2 | NP_071784.1 | 5, 6 | 848 | ||
XM_006230892.2 | XP_006230954.1 | 4, 5 | 596 | |||
XM_006230893.2 | XP_006230955.1 | 4, 5 | 475 | |||
XM_006230894.2 | XP_006230956.1 | 5, 6 | 495 | |||
XM_008760170.1 | XP_008758392.1 | 5, 6 | 1582 | |||
XM_008760171.1 | XP_008758393.1 | 4, 5 | 1243 | |||
XM_008760172.1 | XP_008758394.1 | 5, 6 | 1142 | |||
XM_008760173.1 | XP_008758395.1 | 4, 5 | 1122 | |||
GenBank mRNA | AB016800.1 | - | 3, 4 | 539 | ||
AF071500.1 | - | 4, 5 | 451 | |||
AF272393.1 | - | 5, 6 | 848 | |||
BC081688.1 | - | 3, 4 | 539 | 7109221 |
