siRNA Details
Gene Symbol: Gng11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022396.2 | NP_071791.1 | 1 | 371 | ||
GenBank mRNA | AF257110.1 | - | 1 | 92 | ||
BC097320.1 | - | 1 | 371 | 7455035 |
Gene Symbol: LOC100912034 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749698.3 | XP_003749746.1 | 1 | 529 |

More Information
Other Species Targeted

Gene Ontology Categories:
