siRNA Details
Gene Symbol: Acads | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022512.2 | NP_071957.1 | 5 | 585 | ||
GenBank mRNA | BC072545.1 | - | 5 | 604 | 7105799 | |
J05030.1 | - | 5 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
