siRNA Details
Gene Symbol: Rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022514.1 | NP_071959.1 | 3, 4 | 381 | ||
GenBank mRNA | BC058474.1 | - | 4, 5 | 411 | 6919338 | |
BC091566.1 | - | 4, 5 | 379 | 7388530 | ||
FQ210011.1 | - | 4, 5 | 397 | |||
FQ214023.1 | - | 4, 5 | 401 | |||
FQ217713.1 | - | 3, 4 | 321 | |||
FQ221292.1 | - | 4, 5 | 406 | |||
FQ221643.1 | - | 4, 5 | 398 | |||
FQ221684.1 | - | 4, 5 | 399 | |||
FQ221938.1 | - | 4, 5 | 412 | |||
FQ222018.1 | - | 4, 5 | 401 | |||
FQ222606.1 | - | 4, 5 | 402 | |||
FQ222991.1 | - | 4, 5 | 403 | |||
FQ223942.1 | - | 3, 4 | 330 | |||
FQ224664.1 | - | 4, 5 | 399 | |||
FQ228505.1 | - | 4, 5 | 398 | |||
FQ228603.1 | - | 4, 5 | 399 | |||
FQ228786.1 | - | 4, 5 | 398 | |||
X07424.1 | - | 3, 4 | 381 |

More Information
Other Species Targeted

Gene Ontology Categories:
