siRNA Details
Gene Symbol: Rpl27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022514.1 | NP_071959.1 | 2 | 165 | ||
GenBank mRNA | BC058474.1 | - | 3 | 195 | 6919338 | |
BC091566.1 | - | 3 | 163 | 7388530 | ||
FQ210011.1 | - | 3 | 181 | |||
FQ214023.1 | - | 3 | 185 | |||
FQ217713.1 | - | 2 | 105 | |||
FQ221292.1 | - | 3 | 190 | |||
FQ221643.1 | - | 3 | 182 | |||
FQ221684.1 | - | 3 | 183 | |||
FQ221938.1 | - | 3 | 196 | |||
FQ222018.1 | - | 3 | 185 | |||
FQ222606.1 | - | 3 | 186 | |||
FQ222991.1 | - | 3 | 187 | |||
FQ223942.1 | - | 2 | 114 | |||
FQ224664.1 | - | 3 | 183 | |||
FQ228505.1 | - | 3 | 182 | |||
FQ228603.1 | - | 3 | 183 | |||
FQ228786.1 | - | 3 | 182 | |||
X07424.1 | - | 2 | 165 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
