siRNA Details
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012906.1 | NP_037038.1 | 7 | 1226 | ||
NM_022520.1 | NP_071965.1 | 7 | 1226 | |||
GenBank mRNA | CB795867.1 | - | 3 | 330 | ||
FM030557.1 | - | 2 | 92 | |||
X77934.1 | - | 7 | 1214 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
