siRNA Details
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012906.1 | NP_037038.1 | 17 | 2355 | ||
NM_022520.1 | NP_071965.1 | 16 | 2319 | |||
GenBank mRNA | FM112636.1 | - | 3 | 261 | ||
M31322.1 | - | 6 | 533 |
