siRNA Details
Gene Symbol: Aplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012906.1 | NP_037038.1 | 10 | 1632 | ||
NM_022520.1 | NP_071965.1 | 10 | 1632 | |||
GenBank mRNA | FM030557.1 | - | 5 | 498 | ||
FM039816.1 | - | 1 | 24 | |||
X77934.1 | - | 10 | 1620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
