siRNA Details
Gene Symbol: Oat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022521.3 | NP_071966.1 | 9 | 1254 | ||
GenBank mRNA | BC061551.1 | - | 9 | 1257 | 6890520 | |
FQ210816.1 | - | 3 | 355 | |||
FQ223550.1 | - | 1 | 14 | |||
M11842.1 | - | 8 | 1197 |
