siRNA Details
Gene Symbol: Dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022526.2 | NP_071971.1 | 2, 3 | 350 | ||
GenBank mRNA | BC060569.1 | - | 2, 3 | 168 | 6890457 | |
FQ218203.1 | - | 2, 3 | 235 | |||
FQ228698.1 | - | 2, 3 | 188 | |||
FQ228865.1 | - | 2, 3 | 194 | |||
U05334.1 | - | 2, 3 | 351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
