siRNA Details
Gene Symbol: Crybb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031690.1 | NP_113878.1 | 2 | 129 | ||
XM_006249572.1 | XP_006249634.1 | 3 | 167 | |||
GenBank mRNA | AF287304.1 | - | 2 | 101 | ||
X05899.1 | - | 2 | 129 |

More Information
Other Species Targeted

Gene Ontology Categories:
