siRNA Details
Gene Symbol: Pdlim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053326.1 | NP_445778.1 | 10 | 1383 | ||
XM_006233404.2 | XP_006233466.1 | 16 | 2619 | |||
XM_006233405.2 | XP_006233467.1 | 15 | 2602 | |||
XM_006233406.2 | XP_006233468.1 | 14 | 2582 | |||
XM_006233407.2 | XP_006233469.1 | 15 | 2548 | |||
XM_006233409.2 | XP_006233471.1 | 16 | 2286 | |||
XM_006233410.2 | XP_006233472.1 | 15 | 2270 | |||
XM_006233411.2 | XP_006233473.1 | 14 | 2250 | |||
XM_006233412.2 | XP_006233474.1 | 13 | 2210 | |||
XM_006233413.2 | XP_006233475.1 | 16 | 2212 | |||
XM_006233414.2 | XP_006233476.1 | 14 | 2200 | |||
XM_006233415.2 | XP_006233477.1 | 13 | 2181 | |||
XM_006233416.2 | XP_006233478.1 | 11 | 2176 | |||
XM_006233418.2 | XP_006233480.1 | 11 | 1617 | |||
XM_006233419.2 | XP_006233481.1 | 10 | 1597 | |||
GenBank mRNA | U48247.1 | - | 10 | 1383 |
