siRNA Details
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023954.1 | NP_076444.1 | 1 | 2341 | ||
GenBank mRNA | AJ276137.1 | - | 1 | 184 | ||
X17621.1 | - | 1 | 1125 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kcna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023954.1 | NP_076444.1 | 1 | 2341 | ||
GenBank mRNA | AJ276137.1 | - | 1 | 184 | ||
X17621.1 | - | 1 | 1125 |