siRNA Details
Gene Symbol: Calu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033898.1 | NP_001029070.1 | 3 | 394 | ||
GenBank mRNA | AJ001929.1 | - | 3 | 427 | ||
BC101908.1 | - | 3 | 394 | 7933832 | ||
FQ222101.1 | - | 2 | 237 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_184053, AK147756, AY225334, AK166705.
Gene Ontology Categories:
