siRNA Details
Gene Symbol: Ppib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022536.2 | NP_071981.1 | 1 | 121 | ||
GenBank mRNA | AF071225.1 | - | 1 | 121 | ||
BC061971.1 | - | 1 | 150 | 5623342 | ||
FQ211358.1 | - | 1 | 125 | |||
FQ212031.1 | - | 1 | 150 | |||
FQ219792.1 | - | 1 | 159 | |||
FQ219926.1 | - | 1 | 171 | |||
FQ221020.1 | - | 1 | 174 | |||
FQ221248.1 | - | 1 | 169 | |||
FQ222696.1 | - | 1 | 283 | |||
FQ223162.1 | - | 1 | 168 | |||
FQ228087.1 | - | 1 | 164 | |||
FQ229174.1 | - | 1 | 171 | |||
FQ229214.1 | - | 1 | 165 | |||
FQ230004.1 | - | 175 | ||||
FQ231656.1 | - | 1 | 164 | |||
FQ232077.1 | - | 1 | 160 | |||
FQ232920.1 | - | 1 | 174 |

More Information
Other Species Targeted

Gene Ontology Categories:
