siRNA Details
Gene Symbol: Ppap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022538.2 | NP_071983.2 | 6 | 841 | ||
XM_006231949.2 | XP_006232011.1 | 6 | 1209 | |||
GenBank mRNA | AF503609.1 | - | 6 | 747 | ||
AF503610.1 | - | 5 | 592 | |||
BC061815.1 | - | 6 | 841 | 5598693 | ||
FQ229687.1 | - | 4 | 436 | |||
JQ013728.1 | - | 6 | 779 | |||
U90556.1 | - | 6 | 756 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
