siRNA Details
Gene Symbol: Ppap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022538.2 | NP_071983.2 | 2, 3 | 299 | ||
GenBank mRNA | BC061815.1 | - | 2, 3 | 299 | 5598693 | |
JQ013728.1 | - | 2, 3 | 237 | |||
U90556.1 | - | 2, 3 | 214 |

More Information
Other Species Targeted

Gene Ontology Categories:
