siRNA Details
Gene Symbol: Ccdc80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022543.2 | NP_071988.2 | 5 | 2614 | ||
GenBank mRNA | AF223677.1 | - | 5 | 2721 | ||
AY548105.1 | - | 5 | 2614 | |||
BC070926.1 | - | 4 | 1952 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
