siRNA Details
Gene Symbol: Ndrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271091.1 | NP_001258020.1 | 10, 11 | 895 | ||
NM_001271092.1 | NP_001258021.1 | 10, 11 | 895 | |||
NM_001271093.1 | NP_001258022.1 | 10, 11 | 842 | |||
NM_001271094.1 | NP_001258023.1 | 10, 11 | 842 | |||
NM_001271095.1 | NP_001258024.1 | 8, 9 | 760 | |||
NM_031967.2 | NP_114173.2 | 8, 9 | 760 | |||
XM_006255123.2 | XP_006255185.1 | 10, 11 | 837 | |||
GenBank mRNA | AF045564.1 | - | 8, 9 | 692 | ||
AY217029.1 | - | 8, 9 | 731 | |||
AY217030.1 | - | 10, 11 | 842 | |||
AY217031.1 | - | 10, 11 | 842 | |||
AY217032.1 | - | 10, 11 | 890 | |||
AY217033.1 | - | 10, 11 | 890 | |||
AY255791.1 | - | 8, 9 | 731 | |||
CB585874.1 | - | 4, 5 | 317 | |||
CB713901.1 | - | 1, 2 | 75 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
