siRNA Details
Gene Symbol: Scrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033499.1 | NP_277034.1 | 2 | 151 | ||
GenBank mRNA | AJ132434.1 | - | 2 | 151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
