siRNA Details
Gene Symbol: Masp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172043.1 | NP_742040.1 | 11 | 2051 | ||
GenBank mRNA | AJ277747.1 | - | 10 | 1993 | ||
BC128700.1 | - | 11 | 2052 | 7324989 | ||
Y18564.1 | - | 10 | 1830 | |||
Y18565.1 | - | 4 | 976 | |||
Y18566.1 | - | 8 | 1583 | |||
Y18571.1 | - | 5 | 1058 | |||
Y18572.1 | - | 10 | 1888 |
