siRNA Details
Gene Symbol: Decr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171996.3 | NP_741993.1 | 4 | 415 | ||
XM_006246024.2 | XP_006246086.1 | 5 | 615 | |||
GenBank mRNA | AF044574.1 | - | 4 | 337 | ||
BC070959.1 | - | 4 | 416 | 7097883 | ||
FQ218511.1 | - | 4 | 390 |
Gene Symbol: Rab11fip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY383698.1 | - | 679 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
