siRNA Details
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172033.2 | NP_742030.1 | 7 | 694 | ||
XM_006229787.2 | XP_006229849.1 | 8 | 932 | |||
XM_006229788.2 | XP_006229850.1 | 7 | 1093 | |||
XM_006229789.2 | XP_006229851.1 | 8 | 1003 | |||
XM_006229791.2 | XP_006229853.1 | 6 | 1460 | |||
XM_008759795.1 | XP_008758017.1 | 6 | 984 | |||
GenBank mRNA | AF081582.1 | - | 7 | 694 | ||
AF118562.1 | - | 7 | 664 | |||
BC078704.1 | - | 6 | 619 | 7124581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
