siRNA Details
Gene Symbol: Hdlbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172039.2 | NP_742036.2 | 19 | 2642 | ||
XM_006245548.1 | XP_006245610.1 | 19 | 2682 | |||
GenBank mRNA | BC105748.1 | - | 19 | 2642 | 5598374 | |
U90725.1 | - | 19 | 2642 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
