siRNA Details
Gene Symbol: Hdlbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172039.2 | NP_742036.2 | 26 | 3741 | ||
XM_006245548.1 | XP_006245610.1 | 26 | 3781 | |||
GenBank mRNA | BC105748.1 | - | 26 | 3741 | 5598374 | |
FQ231933.1 | - | 2 | 250 | |||
U90725.1 | - | 26 | 3741 |

More Information
Other Species Targeted

Gene Ontology Categories:
